Thermo Scientific Chemicals

2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: H27024.03
1 g, Each
Thermo Scientific Chemicals

2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: H27024.03
1 g, Each
Quantity
Catalog number: H27024.03
also known as H27024-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13331-19-6
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Melting Point (clear melt)
102.0-108.0?C
Form
Powder

Description

2-n-propylresorcinol is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-propylresorcinol is a pharmaceutical intermediate.

Solubility
Solightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text