2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals
2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H27024.06
also known as H27024-06
Price (USD)/ Each
244.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
244.00
Add to cart
2-n-Propylresorcinol, 98%, Thermo Scientific Chemicals
Catalog numberH27024.06
Price (USD)/ Each
244.00
-
Add to cart
Chemical Identifiers
CAS13331-19-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥97.5%
Melting Point (clear melt)102.0-108.0?C
FormPowder
2-n-propylresorcinol is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-propylresorcinol is a pharmaceutical intermediate.

Solubility
Solightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. H Dressler. The Properties and Chemistry of Resorcinol. Resorcinol. 1994 , volume 5-25.
  2. M. A. Stealeya,; R. L. Shonea,; M. Miyanoa. A Convenient Synthesis of 2-Alkyl-mono-O-alkylated and 2-Alkyl Resorcinols. An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1990, 12(20), pg no.