1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H27036.03
also known as H27036-03
Price (USD)/ Each
192.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
192.00
Add to cart
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH27036.03
Price (USD)/ Each
192.00
-
Add to cart
Chemical Identifiers
CAS903550-26-5
IUPAC Name1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC14H23BN2O3
InChI KeyZZRFDLHBMBHJTI-UHFFFAOYNA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=NN1C1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder
Assay (GC)≥94.0%
Melting Point (clear melt)69-77?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only