Thermo Scientific Chemicals

2-(Diphenylphosphino)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: H27070.09
10 g, Each
Thermo Scientific Chemicals

2-(Diphenylphosphino)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: H27070.09
10 g, Each
Quantity
Catalog number: H27070.09
also known as H27070-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4848-43-5
IUPAC Name
(2-aminoethyl)diphenylphosphane
Molecular Formula
C14H16NP
InChI Key
RXEPBCWNHKZECN-UHFFFAOYSA-N
SMILES
NCCP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0% (UK sourced material)
Assay from Supplier's CofA
≥94.0% (GC, US sourced material)
Assay from Supplier's CofA
≥94.0% (P31-NMR, US sourced material)

Description

2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text