Thermo Scientific Chemicals

1-Boc-3-fluoro-4-pyrrolidinone, Thermo Scientific Chemicals

Catalog number: H27082.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-3-fluoro-4-pyrrolidinone, Thermo Scientific Chemicals

Catalog number: H27082.03
1 g, Each
Quantity
Catalog number: H27082.03
also known as H27082-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
845894-03-3
IUPAC Name
tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
Molecular Formula
C9H14FNO3
InChI Key
HPHVNLXMQXEDFX-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CC(F)C(=O)C1
Form
Powder
Purity (DSC)
≥95.0 mol%
Melting Point (clear melt)
75.5-87.5?C
Appearance (Color)
White to pale brown

Description

1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text