1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only