Thermo Scientific Chemicals

Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals

Catalog number: H27092.09
10 g, Each
Thermo Scientific Chemicals

Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals

Catalog number: H27092.09
10 g, Each
Quantity
Catalog number: H27092.09
also known as H27092-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1624-01-7
Appearance (Color)
Clear colorless
Form
Liquid or low-melting solid (clear liquid as melt)
Assay (GC)
≥98.5%
Proton NMR
Conforms to structure

Description

Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text