Thermo Scientific Chemicals

4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27097.03
1 g, Each
Thermo Scientific Chemicals

4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27097.03
1 g, Each
Quantity
Catalog number: H27097.03
also known as H27097-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
165881-36-7
IUPAC Name
4-(4-bromobutyl)-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0²,⁶]decane
Molecular Formula
C14H24BBrO2
InChI Key
DQGAIMVYOVPBSD-UHFFFAOYNA-N
SMILES
CC1(C)C2CC1C1(C)OB(CCCCBr)OC1C2
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0%
Proton NMR
Conforms to structure
Optical Rotation
19.0 ± 0.1? (C=1 in chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text