Thermo Scientific Chemicals

D-Cysteine hydrochloride monohydrate, 99%

Catalog number: H27107.06
5 g, Each
Thermo Scientific Chemicals

D-Cysteine hydrochloride monohydrate, 99%

Catalog number: H27107.06
5 g, Each
Quantity
Catalog number: H27107.06
also known as H27107-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207121-46-8
IUPAC Name
2-amino-3-sulfanylpropanoic acid hydrate hydrochloride
Molecular Formula
C3H10ClNO3S
InChI Key
QIJRTFXNRTXDIP-UHFFFAOYNA-N
SMILES
O.Cl.NC(CS)C(O)=O
Form
Crystals or powder or crystalline powder
Optical Rotation
-3.5° to - 7° (c=5 in 5N HCl)
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text