The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Robert Opitza,; Matthias Müllera,; Cédric Reuterb,; Matthias Baronea,; Arne Soickeb,; Yvette Roskec,; Kirill Piotukh. A modular toolkit to inhibit proline-rich motif-mediated protein-protein interactions. Journal name. 2015, 112(16) ,5011-5016.