2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Nalinrut Masomboon,; Chavalit Ratanatamskul,; Ming-Chun Lu. Chemical Oxidation of 2,6-Dimethylaniline in the Fenton Process. Environ. Sci. Technol.,. 2009, 43 (22) ,8629-8634.
- Matthew S. Bryant,; Henry F. Simmons,; Robert E. Harrell,; Jack A. Hinson. 2,6-Dimethylaniline—hemoglobin adducts from lidocaine in humans. Journal name. 1994, 15 (10),2287-2290..