Thermo Scientific Chemicals

1-Chlorophthalazine, 97%, Thermo Scientific Chemicals

Catalog number: H27143.09
10 g, Each
Thermo Scientific Chemicals

1-Chlorophthalazine, 97%, Thermo Scientific Chemicals

Catalog number: H27143.09
10 g, Each
Quantity
Catalog number: H27143.09
also known as H27143-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5784-45-2
IUPAC Name
1-chlorophthalazine
Molecular Formula
C8H5ClN2
InChI Key
UCOVESIAFFGEOR-UHFFFAOYSA-N
SMILES
ClC1=C2C=CC=CC2=CN=N1
Appearance (Color)
Pale yellow to yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
108-116?C

Description

1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text