Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: H27159.03
1 g, Each
Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog number: H27159.03
1 g, Each
Quantity
Catalog number: H27159.03
also known as H27159-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1486-28-8
IUPAC Name
methyldiphenylphosphane
Molecular Formula
C13H13P
InChI Key
UJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILES
CP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.6225-1.6275 @ 20°C

Description

Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text