5-Fluoro-1,3-dimethyl-2-nitrobenzene, 98%, Thermo Scientific Chemicals
5-Fluoro-1,3-dimethyl-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-1,3-dimethyl-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27166.03
also known as H27166-03
Price (USD)/ Each
79.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
79.20
Add to cart
5-Fluoro-1,3-dimethyl-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Catalog numberH27166.03
Price (USD)/ Each
79.20
-
Add to cart
Chemical Identifiers
CAS315-12-8
IUPAC Name5-fluoro-1,3-dimethyl-2-nitrobenzene
Molecular FormulaC8H8FNO2
InChI KeyKDPQDERPJHLWGF-UHFFFAOYSA-N
SMILESCC1=CC(F)=CC(C)=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)51.5-57.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only