6-Nitrochromone, 97%, Thermo Scientific Chemicals
6-Nitrochromone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Nitrochromone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H27167.14
also known as H27167-14
Price (USD)/ Each
227.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
227.00
Add to cart
6-Nitrochromone, 97%, Thermo Scientific Chemicals
Catalog numberH27167.14
Price (USD)/ Each
227.00
-
Add to cart
Chemical Identifiers
CAS51484-05-0
IUPAC Name6-nitro-4H-chromen-4-one
Molecular FormulaC9H5NO4
InChI KeyORWADBVBOPTYQT-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2OC=CC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)169.5-178.5?C
Appearance (Color)White to yellow or pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only