4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals
4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H27174.14
also known as H27174-14
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals
Catalog numberH27174.14
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS103204-69-9
IUPAC Name4-acetamido-2-methylbenzoic acid
Molecular FormulaC10H11NO3
InChI KeyAQPDTYYKDYMCTH-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC(C)=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
FormPowder
Assay (HPLC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only