Thermo Scientific Chemicals

1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, 98%, Thermo Scientific Chemicals

Catalog number: H27177.06
5 g, Each
Thermo Scientific Chemicals

1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, 98%, Thermo Scientific Chemicals

Catalog number: H27177.06
5 g, Each
Quantity
Catalog number: H27177.06
also known as H27177-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
223437-11-4
IUPAC Name
1-butyl-1-methylpyrrolidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular Formula
C11H20F6N2O4S2
InChI Key
HSLXOARVFIWOQF-UHFFFAOYSA-N
SMILES
CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Appearance (Color)
Clear colorless to orange
Assay by NMR
≥97.5%
Water Content (Karl Fischer Titration)
≤100ppm
Proton NMR
Conforms to structure
Form
Liquid

Description

1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

Solubility
Miscible with methanol, acetonitrile, acetone, dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text