Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: H27178.06
5 g, Each
Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: H27178.06
5 g, Each
Quantity
Catalog number: H27178.06
also known as H27178-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
171058-17-6
IUPAC Name
3-hexyl-1-methyl-1H-imidazol-3-ium
Molecular Formula
C10H19N2
InChI Key
RVEJOWGVUQQIIZ-UHFFFAOYSA-N
SMILES
CCCCCC[N+]1=CN(C)C=C1
Identification (FTIR)
Conforms
Refractive Index
1.5135-1.5175 @ 20?C
Appearance (Color)
Clear yellow to orange
Form
Liquid or viscous liquid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text