2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27179.03
also known as H27179-03
Price (USD)/ Each
281.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
281.00
Add to cart
2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH27179.03
Price (USD)/ Each
281.00
-
Add to cart
Chemical Identifiers
CAS458532-86-0
IUPAC Name2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular FormulaC11H15BFNO2
InChI KeyPCLMNCBIXQQRMB-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥94.0%
Melting Point (clear melt)44.5-53.5?C
Used in pharmaceuticals, agrochemicals, and high-tech materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceuticals, agrochemicals, and high-tech materials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Matteson, D. S. Chiral alpha-Haloalkylboronic esters as reagents for asymmetric synthesis. J. Org. Chem. 2013, 78(20),10009–-10023.