Thermo Scientific Chemicals

2-Hydroxycyclohexanone dimer, 90+%, Thermo Scientific Chemicals

Catalog number: H27199.14
25 g, Each
Thermo Scientific Chemicals

2-Hydroxycyclohexanone dimer, 90+%, Thermo Scientific Chemicals

Catalog number: H27199.14
25 g, Each
Quantity
Catalog number: H27199.14
also known as H27199-14
Price (TWD)

Chemical Identifiers

CAS
30282-14-5
IUPAC Name
2-hydroxycyclohexan-1-one
Molecular Formula
C6H10O2
InChI Key
ODZTXUXIYGJLMC-UHFFFAOYNA-N
SMILES
OC1CCCCC1=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (GC)
≥90.0%
Melting Point (clear melt)
92.0-105.0?C

Description

It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

Solubility
Partly miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text