Thermo Scientific Chemicals

3-(Methylthio)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27200.14
25 g, Each
Thermo Scientific Chemicals

3-(Methylthio)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27200.14
25 g, Each
Quantity
Catalog number: H27200.14
also known as H27200-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
825-99-0
IUPAC Name
3-(methylsulfanyl)benzoic acid
Molecular Formula
C8H8O2S
InChI Key
PZGADOOBMVLBJE-UHFFFAOYSA-N
SMILES
CSC1=CC=CC(=C1)C(O)=O
Assay (Silylated GC)
≥96.0%
Melting Point (clear melt)
124.5-133.5?C
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text