2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals
2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H27212.06
also known as H27212-06
Price (USD)/ Each
149.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
149.00
Add to cart
2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals
Catalog numberH27212.06
Price (USD)/ Each
149.00
-
Add to cart
Chemical Identifiers
CAS32042-38-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.3040-1.3090 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only