2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals
2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number H27218.09
also known as H27218-09
Price (USD)/ Each
82.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
82.00
Add to cart
2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals
Catalog numberH27218.09
Price (USD)/ Each
82.00
-
Add to cart
Chemical Identifiers
CAS773-99-9
IUPAC Name2-(naphthalen-1-yl)ethan-1-ol
Molecular FormulaC12H12O
InChI KeyRXWNCMHRJCOWDK-UHFFFAOYSA-N
SMILESOCCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-68.0?C
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only