Thermo Scientific Chemicals

Uracil-5-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27219.MD
250 mg, Each
Thermo Scientific Chemicals

Uracil-5-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: H27219.MD
250 mg, Each
Quantity
Catalog number: H27219.MD
also known as H27219-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70523-22-7
IUPAC Name
(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
Molecular Formula
C4H5BN2O4
InChI Key
PVEJOCQTIVCDNO-UHFFFAOYSA-N
SMILES
OB(O)C1=CNC(=O)NC1=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥94.0%
Proton NMR
Conforms to structure

Description

It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text