Thermo Scientific Chemicals

L-m-Tyrosine, 97+%, Thermo Scientific Chemicals

Catalog number: H27224.03
1 g, Each
Thermo Scientific Chemicals

L-m-Tyrosine, 97+%, Thermo Scientific Chemicals

Catalog number: H27224.03
1 g, Each
Quantity
Catalog number: H27224.03
also known as H27224-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
587-33-7
IUPAC Name
(2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
Molecular Formula
C9H11NO3
InChI Key
JZKXXXDKRQWDET-SVGMAFHSNA-N
SMILES
N[C@@H](CC1=CC=CC(O)=C1)C(O)=O
Appearance (Color)
White to cream to brown
Form
Powder
Assay (HPLC)
≥97.0%
Optical Rotation
-9.6 ± 0.3? (C=4 in 1M HCl)

Description

An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, dilute acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text