Thermo Scientific Chemicals

2-Methyl-1-butene, 98%, Thermo Scientific Chemicals

Catalog number: H27230.06
5 g, Each
Thermo Scientific Chemicals

2-Methyl-1-butene, 98%, Thermo Scientific Chemicals

Catalog number: H27230.06
5 g, Each
Quantity
Catalog number: H27230.06
also known as H27230-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-46-2
IUPAC Name
2-methylbut-1-ene
Molecular Formula
C5H10
InChI Key
MHNNAWXXUZQSNM-UHFFFAOYSA-N
SMILES
CCC(C)=C
Form
Liquid
Comment
Purchased in U.K. and U.S.
Assay (GC)
≥97.5% (U.K. specification)
Appearance (Color)
Clear colorless to pale yellow
Assay from Suppliers CofA
≥97.5% (U.S. specification)

Description

2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

Solubility
Soluble in ether, ethanol and benzene. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text