Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals
Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27244.06
also known as H27244-06
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.10 
Save 6.45 (16%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.10 
Save 6.45 (16%)
Add to cart
Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals
Catalog numberH27244.06
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.10 
Save 6.45 (16%)
-
Add to cart
Chemical Identifiers
CAS932750-29-3
IUPAC Nameethyl 2-cyano-3-ethoxybut-2-enoate
Molecular FormulaC9H13NO3
InChI KeyYKXFOGAYPIPTKF-UHFFFAOYSA-N
SMILESCCOC(C)=C(C#N)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)69.5-75.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only