4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals
4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H27247.03
also known as H27247-03
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
Add to cart
4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals
Catalog numberH27247.03
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10386-28-4
IUPAC Name3-iodopyridine-4-carbonitrile
Molecular FormulaC6H3IN2
InChI KeyKMFIQUYSWTUCGD-UHFFFAOYSA-N
SMILESIC1=C(C=CN=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Cream to pale yellow to pale brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only