Thermo Scientific Chemicals

(6-Chloro-2-pyridyl)hydrazine, 95%, Thermo Scientific Chemicals

Catalog number: H27249.03
1 g, Each
Thermo Scientific Chemicals

(6-Chloro-2-pyridyl)hydrazine, 95%, Thermo Scientific Chemicals

Catalog number: H27249.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H27249.03
also known as H27249-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5193-03-3
IUPAC Name
2-chloro-6-hydrazinylpyridine
Molecular Formula
C5H6ClN3
InChI Key
HFTSPKIAXHZROA-UHFFFAOYSA-N
SMILES
NNC1=CC=CC(Cl)=N1
Appearance (Color)
Pale cream to cream
Assay (GC)
≥94.0%
Form
Powder
Melting Point (clear melt)
112.0-125.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text