Dimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Temperature sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, water, moisture, mercury, chlorine, copper and copper compounds.
RUO – Research Use Only
General References:
- João Almeida et. al. Molecular understanding of Sulfuricacid-amine particle nucleation in the atmosphere. Nature. 2013, 502 7471,359-363.
- Siegfried Schobesberger et. al. Molecular understanding of atmospheric particle formation from sulfuric acid and large oxidized organic molecules.. PNAS. 2013, 110(43), 17223-17228 .