Thermo Scientific Chemicals

Dimethylamine, 2M in methanol, Thermo Scientific Chemicals

Catalog number: H27261.AE
100 mL, Each
Thermo Scientific Chemicals

Dimethylamine, 2M in methanol, Thermo Scientific Chemicals

Catalog number: H27261.AE
100 mL, Each
Quantity
Catalog number: H27261.AE
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-40-3
IUPAC Name
dimethylamine
Molecular Formula
C2H7N
InChI Key
ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES
CNC
Appearance (Color)
Clear, colourless
Assay (Non-aqueous acid-base Titration)
1.90 - 2.20M
Form
liquid
Refractive Index
1.3335 - 1.3365 @20?C

Description

Dimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Temperature sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, water, moisture, mercury, chlorine, copper and copper compounds.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text