Thermo Scientific Chemicals

6-Methoxypyridine-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H27263.03
1 g, Each
Thermo Scientific Chemicals

6-Methoxypyridine-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H27263.03
1 g, Each
Quantity
Catalog number: H27263.03
also known as H27263-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1034297-69-2
IUPAC Name
2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C12H18BNO3
InChI Key
VYWCRVNLPOVYJH-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=N1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
White
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or chunks
Melting Point (clear melt)
65.0-74.0?C

Description

It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text