It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Mihaela Plesescu, et al. Synthesis of isotope-labeled HSP90 inhibitor: [13CD3] and [14C]-TAK-459.J. Labelled Compd. Radiopharm.,2014,757(9), 574-578.
Christopher M. McBride, et al. Design, Structure-Activity Relationship, and in Vivo Characterization of the Development Candidate NVP-HSP990.J. Med. Chem.,2014,57(21), 9124-9129.