1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27264.MD
also known as H27264-MD
Price (USD)/ Each
151.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
151.00
Add to cart
1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH27264.MD
Price (USD)/ Each
151.00
-
Add to cart
Chemical Identifiers
CAS68781-13-5
IUPAC Name1-aminocyclopropane-1-carboxylic acid hydrochloride
Molecular FormulaC4H8ClNO2
InChI KeyWTZRHZRMLYNBRM-UHFFFAOYSA-N
SMILESCl.NC1(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only