Tetra-n-butylammonium dihydrogentrifluoride, tech. 90%, Thermo Scientific Chemicals
Tetra-n-butylammonium dihydrogentrifluoride, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butylammonium dihydrogentrifluoride, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H27266.06
also known as H27266-06
Price (USD)/ Each
156.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
156.00
Add to cart
Tetra-n-butylammonium dihydrogentrifluoride, tech. 90%, Thermo Scientific Chemicals
Catalog numberH27266.06
Price (USD)/ Each
156.00
-
Add to cart
Chemical Identifiers
CAS99337-56-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Water Content (Karl Fischer Titration)<2.0%
Refractive Index1.4400-1.4490 @ 20?C
FormViscous liquid
Assay (Non-aqueous acid-base Titration)>88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only