1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H27288.06
also known as H27288-06
Price (USD)/ Each
381.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
381.00
Add to cart
1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH27288.06
Price (USD)/ Each
381.00
-
Add to cart
Chemical Identifiers
CAS127946-77-4
IUPAC Name1-aminocyclopropane-1-carbonitrile hydrochloride
Molecular FormulaC4H7ClN2
InChI KeyPCEIEQLJYDMRFZ-UHFFFAOYSA-N
SMILESCl.NC1(CC1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)Dark cream to cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only