It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
- Carmen Moser, et. al. The homologous series of 1,1'-ferrocenylenebisdihalophosphanes (C5H4PX2)2Fe (X = F, Cl, Br, I): precursors for the first metallocene bridged bisphosphaalkene.Dalton Trans.,2006,732(7), 3879-3885.
- Xingao Peng, et al. Ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation.Tetrahedron Lett.,2008,49(33), 4862-4864.