(R)-1-(Boc-amino)-2-propanol, 98%, Thermo Scientific Chemicals
(R)-1-(Boc-amino)-2-propanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-1-(Boc-amino)-2-propanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H27296.06
also known as H27296-06
Price (USD)/ Each
331.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
331.00
Add to cart
(R)-1-(Boc-amino)-2-propanol, 98%, Thermo Scientific Chemicals
Catalog numberH27296.06
Price (USD)/ Each
331.00
-
Add to cart
Chemical Identifiers
CAS119768-44-4
IUPAC Nametert-butyl N-[(2R)-2-hydroxypropyl]carbamate
Molecular FormulaC8H17NO3
InChI KeyYNJCFDAODGKHAV-ZCFIWIBFSA-N
SMILESC[C@@H](O)CNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Optical Rotation-21.0 ± 3.5° (C=1 in chloroform)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only