Thermo Scientific Chemicals

Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals

Catalog number: H27307.18
50 g, Each
Thermo Scientific Chemicals

Lithium bis(trifluoromethylsulfonyl)imide, 98+%, Thermo Scientific Chemicals

Catalog number: H27307.18
50 g, Each
Quantity
Catalog number: H27307.18
also known as H27307-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90076-65-6
Total Metal Impurities
(by Ion Chromatography): ≤100 ppm
Appearance (Color)
White
Assay (unspecified)
(by Ion Chromatography): ≥98.0%
Water Content (Karl Fischer Titration)
≤0.5%
Form
Crystals or powder or crystalline powder or chunks

Description

Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.

Solubility
Soluble in water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text