Thermo Scientific Chemicals

Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H27318.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H27318.14
25 g, Each
Quantity
Catalog number: H27318.14
also known as H27318-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71486-53-8
IUPAC Name
hydrogen methyl 4-oxopiperidine-3-carboxylate chloride
Molecular Formula
C7H12ClNO3
InChI Key
NMAACQILAGCQPR-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C1CNCCC1=O
Form
Powder
Assay (Titration ex Chloride)
>94.0 to <106.0%
Proton NMR
Conforms to structure
Water Content (Karl Fischer Titration)
<0.3%
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text