Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals
Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number H27324.AU
also known as H27324-AU
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
Add to cart
Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals
Catalog numberH27324.AU
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-50-3
IUPAC Nametrimethylamine
Molecular FormulaC3H9N
InChI KeyGETQZCLCWQTVFV-UHFFFAOYSA-N
SMILESCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colourless
FormLiquid
Assay (Non-aqueous acid-base Titration)30-36%
Refractive Index1.3615-1.3665 @ 20°C
Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

Solubility
Insoluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, brass, zinc, magnesium, aluminum, mercury, mercury oxides, acid chlorides, acid anhydrides, strong reducing agents.
For Manufacturing and Laboratory Use. Not for beverage use

General References:

  1. David S Wishart et. al. HMDB: a knowledgebase for the human metabolome.. Nucleic Acids Research. 2009, (37), D603-D610 .
  2. E L Barrett,; H S Kwan. Bacterial reduction of trimethylamine oxide. Microbiology. 1995, 39 issue , 131-149 .