Thermo Scientific Chemicals

2-Acetylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27327.06
5 g, Each
Thermo Scientific Chemicals

2-Acetylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H27327.06
5 g, Each
Quantity
Catalog number: H27327.06
also known as H27327-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
308103-40-4
IUPAC Name
(2-acetylphenyl)boronic acid
Molecular Formula
C8H9BO3
InChI Key
ZKAOVABYLXQUTI-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1B(O)O
Identification (FTIR)
Conforms
Assay (HPLC)
≥96.0%
Appearance (Color)
White to yellow
Form
Powder and/or chunks

Description

2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text