Thermo Scientific Chemicals

2,5-Dimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: H27328.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: H27328.14
25 g, Each
Quantity
Catalog number: H27328.14
also known as H27328-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1758-25-4
IUPAC Name
2-(2,5-dimethoxyphenyl)acetic acid
Molecular Formula
C10H12O4
InChI Key
BBZDYQUXRFATHZ-UHFFFAOYSA-N
SMILES
COC1=CC=C(OC)C(CC(O)=O)=C1
Appearance (Color)
White to cream to yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
121.0-128.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text