4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals
4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H27330.MD
also known as H27330-MD
Price (USD)/ Each
91.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
91.00
Add to cart
4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals
Catalog numberH27330.MD
Price (USD)/ Each
91.00
-
Add to cart
Chemical Identifiers
CAS261733-18-0
IUPAC Name4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Molecular FormulaC36H27NOP2
InChI KeyHSWZLYXRAOXOLL-UHFFFAOYSA-N
SMILESN1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow or pale green or green/grey
FormCrystals or powder or crystalline powder
Assay by NMR≥98.0%
Identification (FTIR)Conforms
Melting Point243-259?C (may decompose).
View more
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Cheng, W.; Shang, R.; Yu, H.; Fu, Y. Room-Temperature Decarboxylative Couplings of α-Oxocarboxylates with Aryl Halides by Merging Photoredox with Palladium Catalysis. Chem-Eur. J. 2015, 21 (38), 13191-13195.
  2. Natour, S.; Abu-Reziq, R. Functionalized Magnetic Mesoporous Silica Nanoparticle-Supported Palladium Catalysts for Carbonylative Sonogashira Coupling Reactions of Aryl Iodides. ChemCatChem 2015, 7 (14), 2230-2240.