Thermo Scientific Chemicals

Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals

Catalog number: H27349.0F
2500 mL, Each
Thermo Scientific Chemicals

Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals

Catalog number: H27349.0F
2500 mL, Each
Quantity
Catalog number: H27349.0F
also known as H27349-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5614-37-9
IUPAC Name
methoxycyclopentane
Molecular Formula
C6H12O
InChI Key
SKTCDJAMAYNROS-UHFFFAOYSA-N
SMILES
COC1CCCC1
Form
Liquid
Assay (GC)
≥99.90%
Identification (FTIR)
Conforms
Comment
Stabilized with ca. 50ppm BHT
Appearance (Color)
Clear, colorless

Description

Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

Solubility
Miscible with water, alcohols, ketones and hydrocarbons.

Notes
Store in cool place. Air and light sensitive. Incompatible with oxidizing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text