Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals
Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
500 mL
2500 mL
Catalog number H27349.AU
also known as H27349-AU
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
Add to cart
Cyclopentyl methyl ether, stab., 99.9+%, Thermo Scientific Chemicals
Catalog numberH27349.AU
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5614-37-9
IUPAC Namemethoxycyclopentane
Molecular FormulaC6H12O
InChI KeySKTCDJAMAYNROS-UHFFFAOYSA-N
SMILESCOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥99.90%
Identification (FTIR)Conforms
CommentStabilized with ca. 50ppm BHT
Appearance (Color)Clear, colorless
View more
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

Solubility
Miscible with water, alcohols, ketones and hydrocarbons.

Notes
Store in cool place. Air and light sensitive. Incompatible with oxidizing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
RUO – Research Use Only

General References:

  1. Azzena, U.; Carraro, M.; Mamuye, A. D.; Murgia, I.; Pisano, L.; Zedde, G. Cyclopentyl methyl ether-NH4X: a novel solvent/catalyst system for low impact acetalization reactions. Green Chem. 2015, 17 (6), 3281-3284.
  2. Zhang, H. Liquid-liquid phase equilibria of the quaternary system {water (1) + acrylic acid (2) + acetic acid (3) + cyclopentyl methyl ether (4)}: Measurement, correlation, and comparative study. Fluid Phase Equilib. 2015, 403, 23-29.