2-Chloro-D-phenylalanine, 98%, Thermo Scientific Chemicals
2-Chloro-D-phenylalanine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-D-phenylalanine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27359.MD
also known as H27359-MD
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
2-Chloro-D-phenylalanine, 98%, Thermo Scientific Chemicals
Catalog numberH27359.MD
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS80126-50-7
IUPAC Name2-amino-3-(2-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyCVZZNRXMDCOHBG-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)209.0-215.0?C
Enantiomeric excess≥97.5%
FormPowder
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only