2,4'-Bipyridine, 97%, Thermo Scientific Chemicals
2,4'-Bipyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4'-Bipyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27366.MD
also known as H27366-MD
Price (USD)/ Each
86.30
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
86.30
Add to cart
2,4'-Bipyridine, 97%, Thermo Scientific Chemicals
Catalog numberH27366.MD
Price (USD)/ Each
86.30
-
Add to cart
Chemical Identifiers
CAS581-47-5
IUPAC Name2,4'-bipyridine
Molecular FormulaC10H8N2
InChI KeyRMHQDKYZXJVCME-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)57.0-65.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only