Thermo Scientific Chemicals

Di-tert-butyl N,N-diethylphosphoramidite, 94%, Thermo Scientific Chemicals

Catalog number: H27380.03
1 g, Each
Thermo Scientific Chemicals

Di-tert-butyl N,N-diethylphosphoramidite, 94%, Thermo Scientific Chemicals

Catalog number: H27380.03
1 g, Each
Quantity
Catalog number: H27380.03
also known as H27380-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117924-33-1
IUPAC Name
[bis(tert-butoxy)phosphanyl]diethylamine
Molecular Formula
C12H28NO2P
InChI Key
KUKSUQKELVOKBH-UHFFFAOYSA-N
SMILES
CCN(CC)P(OC(C)(C)C)OC(C)(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥92.0%
Comment
May form small amount of sediment/precipitate upon storage.
Identification (FTIR)
Conforms

Description

A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

Solubility
Hydrolyzes in water. Soluble in chloroform and methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text