Thermo Scientific Chemicals

Potassium 4-methoxyphenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: H27384.03
1 g, Each
Thermo Scientific Chemicals

Potassium 4-methoxyphenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: H27384.03
1 g, Each
Quantity
Catalog number: H27384.03
also known as H27384-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
192863-36-8
IUPAC Name
potassium trifluoro(4-methoxyphenyl)boranuide
Molecular Formula
C7H7BF3KO
InChI Key
XNYMCUFKZHRYID-UHFFFAOYSA-N
SMILES
[K+].COC1=CC=C(C=C1)[B-](F)(F)F
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥94.0%

Description

It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text